Please login to your online account to display your discounted pricing

anti-Fer, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP120089

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP120089

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot No. P16591]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Concentration 1.12mg/mL
Gene ID (Entrez) 2241
Formulation PBS, 30% glycerol
Test Specificity Human. Predicted to react with mouse and canine based on 100% sequence homology.
Gene Accession No. P16591
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit
Antigen Fer
Regulatory Status RUO
Quantity 0.1mL
Cross Reactivity Human