Please login to your online account to display your discounted pricing

anti-Fer, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot No. P16591]
Antigen Fer
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP120089

 
novus biologicals canada
NBP120089
Unlabeled 0.1mL Each for $309.05

NBP120089SS

 
novus biologicals canada
NBP120089SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot No. P16591]
Antigen Fer
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Predicted to react with mouse and canine based on 100% sequence homology.
Gene Accession No. P16591
Gene ID (Entrez) 2241
Formulation PBS, 30% glycerol
Concentration 1.12mg/mL