Please login to your online account to display your discounted pricing

anti-FE65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058360SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058360T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 15% glycerol
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 322
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Primary or Secondary Primary
Host Species Rabbit
Regulatory Status RUO
Immunogen Recombinant rat FE65
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Format Affinity Purified
Antigen FE65