Please login to your online account to display your discounted pricing

anti-FE65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058360

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058360

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 322
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Immunogen Recombinant rat FE65
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Formulation PBS, 15% glycerol
Host Species Rabbit
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Antigen FE65
Regulatory Status RUO