missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FE65, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Recombinant rat FE65
Antigen FE65
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11058360 Novus Biologicals Canada
NB11058360
0.1mL Each for $309.05
NB11058360T Novus Biologicals Canada
NB11058360SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
FE65
Affinity Purified
RUO
322
Recombinant rat FE65
Polyclonal
Primary
Immunoprecipitation,Western Blot
PBS, 15% glycerol