Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), PerCP, Clone: 275003, R&D Systems


Manufacturer: r & d systems  FAB19601C025

 View more versions of this product

Catalog No. FAB19601C02

  • / Each
 

Description & Specifications

Specifications

Antigen Fc gamma RIII (CD16)
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fcγ RIII. Ala31-Thr215
Primary or Secondary Primary
Format Purified
Host Species Rat
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone 275003
Conjugate PerCP
Cross Reactivity Mouse