Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), PerCP, Clone: 245536, R&D Systems


Manufacturer: r & d systems  RDSFAB2546C025

 View more versions of this product

Catalog No. FAB2546C025

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a
Format Purified
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fcγ RIIIA/B (CD16). Thr20-Gln208
Regulatory Status RUO
Host Species Mouse
Quantity 25 Tests
Clone 245536
Conjugate PerCP
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen Fc gamma RIII (CD16)
Purification Method Protein A or G purified from hybridoma culture supernatant