Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), PerCP, Clone: 245536, R&D Systems


Manufacturer: r & d systems  RDSFAB2546C100

 View more versions of this product

Catalog No. FAB2546C100

  • / Each
 

Description & Specifications

Specifications

Antigen Fc gamma RIII (CD16)
Format Purified
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fcγ RIIIA/B (CD16). Thr20-Gln208
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 245536
Conjugate PerCP
Cross Reactivity Human