Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), Fluorescein, Clone: 245536, R&D Systems


Manufacturer: r & d systems  RDSFAB2546F025

 View more versions of this product

Catalog No. FAB2546F025

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Clone 245536
Cross Reactivity Human
Antigen Fc gamma RIII (CD16)
Format Purified
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fcγ RIIIA/B (CD16). Thr20-Gln208
Isotype IgG2a
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Fluorescein
Purification Method Protein A or G purified from hybridoma culture supernatant