Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), APC, Clone: 245536, R&D Systems


Manufacturer: r & d systems  RDSFAB2546A100

 View more versions of this product

Catalog No. FAB2546A100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG2a
Regulatory Status RUO
Cross Reactivity Human
Quantity 100 Tests
Conjugate APC
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Clone 245536
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fcγ RIIIA/B (CD16). Thr20-Gln208
Antigen Fc gamma RIII (CD16)
Purification Method Protein A or G purified from hybridoma culture supernatant