Please login to your online account to display your discounted pricing

anti-Fc gamma RIII (CD16), APC, Clone: 245536, R&D Systems

$129.05 - $389.76

Specifications

Antigen Fc gamma RIII (CD16)
Clone 245536
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2546A025

 
r & d systems
RDSFAB2546A025
APC 25 Tests Each for $129.05

FAB2546A100

 
r & d systems
RDSFAB2546A100
APC 100 Tests Each for $389.76
Description & Specifications

Specifications

Antigen Fc gamma RIII (CD16)
Clone 245536
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fcγ RIIIA/B (CD16). Thr20-Gln208
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry