Please login to your online account to display your discounted pricing

anti-FBXO44, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein.
Antigen FBXO44
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100467SS

 
novus biologicals canada
NB100467SS
Unlabeled 0.025mL Each for $109.00

NB100467

 
novus biologicals canada
NB100467
Unlabeled 0.1mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein.
Antigen FBXO44
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity FBG3 protein. Human. Cross-reactivity with rat and mouse is likely due to high identity (94%) between the immunogen and these rodent sequences.
Gene Accession No. Q9H4M3
Gene ID (Entrez) 93611
Formulation PBS
Concentration 1mg/mL