Please login to your online account to display your discounted pricing

anti-FBXO44, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100467

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100467

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 93611
Cross Reactivity Human
Gene Accession No. Q9H4M3
Test Specificity FBG3 protein. Human. Cross-reactivity with rat and mouse is likely due to high identity (94%) between the immunogen and these rodent sequences.
Antigen FBXO44
Immunogen A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein.
Concentration 1mg/mL
Primary or Secondary Primary
Quantity 0.1mL
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting