Please login to your online account to display your discounted pricing

anti-FBX09, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human FBXO9 was used as the immunogen for this antibody.
Antigen FBX09
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227364

 
novus biologicals canada
NBP227364
Unlabeled 0.1mg Each for $257.41

NBP227364SS

 
novus biologicals canada
NBP227364SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human FBXO9 was used as the immunogen for this antibody.
Antigen FBX09
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Reacts with human. Predicted to cross react with primates by homology (100% homologous to most ape species and 95% homologous to most monkey species).
Gene Accession No. Q9Uk97
Gene ID (Entrez) 26268
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL