Please login to your online account to display your discounted pricing

anti-FAT4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178381SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178381SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 79633
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q6V0I7
Conjugate Unlabeled
Concentration 0.68mg/mL
Formulation PBS, 30% glycerol
Immunogen A synthetic peptide made to a C-terminal portion of the human FAT4 protein (between residues 4000-4500) [UniProt Q6V0I7]
Format Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen FAT4