missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-FAT4, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human FAT4 protein (between residues 4000-4500) [UniProt Q6V0I7]
Antigen FAT4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178381 Novus Biologicals Canada
NBP178381
0.1mL Each for $309.05
NBP178381SS Novus Biologicals Canada
NBP178381SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
FAT4
Affinity Purified
RUO
Q6V0I7
PBS, 30% glycerol
A synthetic peptide made to a C-terminal portion of the human FAT4 protein (between residues 4000-4500) [UniProt Q6V0I7]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
79633
0.68mg/mL