Please login to your online account to display your discounted pricing

anti-FANCM, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11098674SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11098674T

  • / Each
 

Description & Specifications

Specifications

Antigen FANCM
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q8IYD8
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCM (within residues 1100-1200). [Swiss-Prot No. Q8IYD8]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57697
Target Species Human