Please login to your online account to display your discounted pricing

anti-FANCM, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11098674

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11098674

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q8IYD8
Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57697
Quantity 0.1mL
Cross Reactivity Human
Format Affinity Purified
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Immunogen Synthetic peptide made to an internal portion of human FANCM (within residues 1100-1200). [Swiss-Prot No. Q8IYD8]
Antigen FANCM