Please login to your online account to display your discounted pricing

anti-FANCM, Polyclonal, Novus Biologicals™

$109.00 - $321.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCM (within residues 1100-1200). [Swiss-Prot No. Q8IYD8]
Antigen FANCM
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11098674

 
novus biologicals canada
NB11098674
Unlabeled 0.1mL Each for $321.00

NB11098674T

 
novus biologicals canada
NB11098674SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCM (within residues 1100-1200). [Swiss-Prot No. Q8IYD8]
Antigen FANCM
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q8IYD8
Gene ID (Entrez) 57697
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL