Please login to your online account to display your discounted pricing

anti-FANCG, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of human FANCG (within amino acids 550-622). [SwissProt No. O15287]
Antigen FANCG
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002566SS

 
novus biologicals canada
NB1002566SS
Unlabeled 0.025mL Each for $139.00

NB1002566

 
novus biologicals canada
NB1002566
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of human FANCG (within amino acids 550-622). [SwissProt No. O15287]
Antigen FANCG
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Human. Immunogen sequence has 88% homology to mouse.
Gene Accession No. O15287
Gene ID (Entrez) 2189