Please login to your online account to display your discounted pricing

anti-FANCD2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118976

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118976

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 2177
Gene Accession No. Q9BXW9
Host Species Rabbit
Regulatory Status RUO
Immunogen N-terminal human FANCD2 fusion protein [Swiss-Prot No.Q9BXW9].
Quantity 0.1mL
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Antigen FANCD2