Please login to your online account to display your discounted pricing

anti-FANCD2, Polyclonal, Novus Biologicals™

$160.00 - $160.00

Specifications

Host Species Rabbit
Immunogen N-terminal human FANCD2 fusion protein [Swiss-Prot No.Q9BXW9].
Antigen FANCD2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118976SS

 
novus biologicals canada
NBP118976SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen N-terminal human FANCD2 fusion protein [Swiss-Prot No.Q9BXW9].
Antigen FANCD2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Gene Accession No. Q9BXW9
Gene ID (Entrez) 2177