Please login to your online account to display your discounted pricing

anti-FANCC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118979SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118979SS

  • / Each
 

Description & Specifications

Specifications

Antigen FANCC
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Antisera
Gene Accession No. Q00597
Host Species Rabbit
Immunogen N-terminal human FANCC protein [Swiss-Prot No.Q00597].
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2176
Target Species Human
Target Species Primate