Please login to your online account to display your discounted pricing

anti-FANCC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118979

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118979

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. Q00597
Host Species Rabbit
Gene ID (Entrez) 2176
Conjugate Unlabeled
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Primate
Immunogen N-terminal human FANCC protein [Swiss-Prot No.Q00597].
Regulatory Status RUO
Quantity 0.1mL
Primary or Secondary Primary
Antigen FANCC