Please login to your online account to display your discounted pricing

anti-FANCC, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen N-terminal human FANCC protein [Swiss-Prot No.Q00597].
Antigen FANCC
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118979

 
novus biologicals canada
NBP118979
Unlabeled 0.1mL Each for $309.05

NBP118979SS

 
novus biologicals canada
NBP118979SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen N-terminal human FANCC protein [Swiss-Prot No.Q00597].
Antigen FANCC
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q00597
Gene ID (Entrez) 2176