Please login to your online account to display your discounted pricing

anti-FAM59A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224573SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224573SS

  • / Each
 

Description & Specifications

Specifications

Antigen FAM59A
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9H706
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human FAM59A was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64762
Test Specificity The IFITM3 amino acid sequence used as immunogen is 100% homologous in mouse and rat and 75% homologous in cow.
Target Species Equine
Target Species Human