Please login to your online account to display your discounted pricing

anti-FAM59A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224573

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224573

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity The IFITM3 amino acid sequence used as immunogen is 100% homologous in mouse and rat and 75% homologous in cow.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 64762
Gene Accession No. Q9H706
Immunogen A portion of amino acids 1-50 of human FAM59A was used as the immunogen for the antibody.
Quantity 0.1mg
Antigen FAM59A
Cross Reactivity Equine
Cross Reactivity Human
Format Affinity Purified
Conjugate Unlabeled
Regulatory Status RUO