Please login to your online account to display your discounted pricing

anti-FAM59A, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human FAM59A was used as the immunogen for the antibody.
Antigen FAM59A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224573

 
novus biologicals canada
NBP224573
Unlabeled 0.1mg Each for $257.41

NBP224573SS

 
novus biologicals canada
NBP224573SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human FAM59A was used as the immunogen for the antibody.
Antigen FAM59A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The IFITM3 amino acid sequence used as immunogen is 100% homologous in mouse and rat and 75% homologous in cow.
Gene Accession No. Q9H706
Gene ID (Entrez) 64762
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL