Please login to your online account to display your discounted pricing

anti-FADD, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP145552SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP145552SS

  • / Each
 

Description & Specifications

Specifications

Antigen FADD
Applications Immunoprecipitation
Applications Western Blot
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation phosphate-buffered saline (PBS) containing 30% glycerol, 0.5% BSA
Gene Accession No. Q13158
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 128 of human FADD.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8772
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat