Please login to your online account to display your discounted pricing

anti-Factor IX Polyclonal, Invitrogen™


Factor IX Polyclonal antibody PA1-74023 from Thermo Scientific™ specifically detects Factor IX in Human samples, and it is validated for ELISA and Immunoelectrophoresis

Manufacturer: pierce biotechnology inc  PA174023

Catalog No. PIPA174023

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunoelectrophoresis
Formulation 200μg purified Antibody in proprietary buffer, with 50% glycerol and no preservative;
Gene Accession No. P00740
Species Reactivity Human
Storage Requirements Avoid Freeze/Thaw Cycles
Gene ID (Entrez) 2158
Gene Symbols F9
Antigen Factor IX
Format Purified
Quantity 200μg
Isotype IgG
Host Species Goat
Conjugate HRP
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Factor IX from human plasma.

Factor IX Polyclonal antibody PA1-74023 from Thermo Scientific™ specifically detects Factor IX in Human samples, and it is validated for ELISA and Immunoelectrophoresis


Host Species: Goat
Cross Reactivity: Human
Isotype: IgG
Immunogen: Factor IX from human plasma.
Formulation: 200μg purified Antibody in proprietary buffer, with 50% glycerol and no preservative;
Uniprot Accession: P00740
Gene Symbol: F9
Entrez GeneID: 2158
PA1-74023 detects Factor IX in Human samples. PA1-74023 has been successfully used in ELISA and Immunoelectrophoresis procedures. PA1-74023 The immunogen is Factor IX from human plasma..