Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118977

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118977

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O15360
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Rabbit
Gene ID (Entrez) 2175
Antigen FACA/FANCA
Quantity 0.1mL
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Immunogen Two synthetic peptides made to a region of human FANCA (within amino acids 930-1100 and 1390-1455) [Swiss Prot No.O15360]
Cross Reactivity Human