Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCA (within residues 800-900). [UniProt No. O15360]
Antigen FACA/FANCA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11098673

 
novus biologicals canada
NB11098673
Unlabeled 0.1mL Each for $255.80

NB11098673T

 
novus biologicals canada
NB11098673SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCA (within residues 800-900). [UniProt No. O15360]
Antigen FACA/FANCA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. O15360
Gene ID (Entrez) 2175
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL