Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™

$139.00 - $321.00

Specifications

Host Species Rabbit
Immunogen Two synthetic peptides made to a region of human FANCA (within amino acids 930-1100 and 1390-1455) [Swiss Prot No.O15360]
Antigen FACA/FANCA
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118977

 
novus biologicals canada
NBP118977
Unlabeled 0.1mL Each for $321.00

NBP118977SS

 
novus biologicals canada
NBP118977SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Two synthetic peptides made to a region of human FANCA (within amino acids 930-1100 and 1390-1455) [Swiss Prot No.O15360]
Antigen FACA/FANCA
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. O15360
Gene ID (Entrez) 2175