Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11098673SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11098673T

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL
Gene Accession No. O15360
Conjugate Unlabeled
Gene ID (Entrez) 2175
Primary or Secondary Primary
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of human FANCA (within residues 800-900). [UniProt No. O15360]
Quantity 0.025mL
Antigen FACA/FANCA
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Format Affinity Purified