Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11098673

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11098673

  • / Each
 

Description & Specifications

Specifications

Antigen FACA/FANCA
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. O15360
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FANCA (within residues 800-900). [UniProt No. O15360]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2175
Target Species Human