Please login to your online account to display your discounted pricing

anti-FACA/FANCA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118977SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118977SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2175
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Gene Accession No. O15360
Quantity 0.025mL
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Antigen FACA/FANCA
Cross Reactivity Human
Host Species Rabbit
Immunogen Two synthetic peptides made to a region of human FANCA (within amino acids 930-1100 and 1390-1455) [Swiss Prot No.O15360]