Please login to your online account to display your discounted pricing

anti-EZH2/KMT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11083976SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11083976T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q15910
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Concentration 1mg/mL
Gene ID (Entrez) 2146
Conjugate Unlabeled
Immunogen Synthetic peptide made to an internal portion of the human KMT6/EZH2 protein (within residues 300-400). [Swiss-Prot No. Q15910]
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mL
Format Affinity Purified
Primary or Secondary Primary
Host Species Rabbit
Antigen EZH2/KMT6