Please login to your online account to display your discounted pricing

anti-EZH2/KMT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11083976

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11083976

  • / Each
 

Description & Specifications

Specifications

Antigen EZH2/KMT6
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q15910
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human KMT6/EZH2 protein (within residues 300-400). [Swiss-Prot No. Q15910]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2146
Target Species Human
Target Species Murine
Target Species Primate