Please login to your online account to display your discounted pricing

anti-Exosome Component 9, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Antigen Exosome Component 9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171702

 
novus biologicals canada
NBP171702
Unlabeled 0.1mL Each for $309.05

NBP171702SS

 
novus biologicals canada
NBP171702SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Antigen Exosome Component 9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity Human, Mouse and Chicken. Immunogen has 83% identity to rat and 90% identity to bovine.
Gene Accession No. Q06265
Gene ID (Entrez) 5393
Formulation PBS, 30% glycerol
Concentration 0.6mg/mL