Please login to your online account to display your discounted pricing

anti-ETS1 associated protein II, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600247SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600247SS

  • / Each
 

Description & Specifications

Specifications

Antigen ETS1 associated protein II
Gene Accession No. O95551
Gene ID (Entrez) 51567
Test Specificity This is specific for EAPII.
Conjugate Unlabeled
Formulation PBS
Primary or Secondary Primary
Format Affinity Purified
Applications Western Blotting
Concentration 1mg/mL
Cross Reactivity Human
Regulatory Status RUO
Immunogen A synthetic peptide made to residues 11-28 (REAAEEEGEPEVKKRRLL) of human EAPII.
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit