Please login to your online account to display your discounted pricing

anti-ETS1 associated protein II, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600247

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600247

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Gene Accession No. O95551
Gene ID (Entrez) 51567
Primary or Secondary Primary
Conjugate Unlabeled
Antigen ETS1 associated protein II
Test Specificity This is specific for EAPII.
Formulation PBS
Quantity 0.1mL
Cross Reactivity Human
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide made to residues 11-28 (REAAEEEGEPEVKKRRLL) of human EAPII.
Applications Western Blotting
Format Affinity Purified
Host Species Rabbit