Please login to your online account to display your discounted pricing

anti-ETS1 associated protein II, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to residues 11-28 (REAAEEEGEPEVKKRRLL) of human EAPII.
Antigen ETS1 associated protein II
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600247

 
novus biologicals canada
NB600247
Unlabeled 0.1mL Each for $309.05

NB600247SS

 
novus biologicals canada
NB600247SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to residues 11-28 (REAAEEEGEPEVKKRRLL) of human EAPII.
Antigen ETS1 associated protein II
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This is specific for EAPII.
Gene Accession No. O95551
Gene ID (Entrez) 51567
Formulation PBS
Concentration 1mg/mL