Please login to your online account to display your discounted pricing

anti-Ets-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056620SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056620T

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene ID (Entrez) 2113
Applications Western Blotting
Formulation 200μL PBS containing 0.2% gelatin
Conjugate Unlabeled
Quantity 0.05mL
Cross Reactivity Human
Antigen Ets-1
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human c-Ets-1.
Format Purified