Please login to your online account to display your discounted pricing

anti-Ets-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056620

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056620

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2113
Monoclonal or Polyclonal Polyclonal
Applications Western Blotting
Primary or Secondary Primary
Formulation 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen Ets-1
Format Purified
Regulatory Status RUO
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human c-Ets-1.
Cross Reactivity Human
Quantity 0.2mL
Host Species Rabbit