Please login to your online account to display your discounted pricing

anti-Ets-1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human c-Ets-1.
Antigen Ets-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056620

 
novus biologicals canada
NB10056620
Unlabeled 0.2mL Each for $255.80

NB10056620T

 
novus biologicals canada
NB10056620SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human c-Ets-1.
Antigen Ets-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 2113
Formulation 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL