Please login to your online account to display your discounted pricing

anti-ERR alpha/NR3B1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP147254SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP147254SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P11474
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Test Specificity Human and mouse. Predicted to react with rat, pig and dog based on 100% sequence homology.
Formulation PBS, 30% glycerol
Conjugate Unlabeled
Host Species Rabbit
Primary or Secondary Primary
Gene ID (Entrez) 2101
Concentration 1mg/mL
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Antigen ERR alpha/NR3B1
Immunogen Synthetic peptide made to an N-terminal portion of human Estrogen Related Receptor alpha (within residues 1-50). [Swiss-Prot No. P11474]
Regulatory Status RUO