missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ERR alpha/NR3B1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human Estrogen Related Receptor alpha (within residues 1-50). [Swiss-Prot No. P11474]
Antigen ERR alpha/NR3B1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP147254 Novus Biologicals Canada
NBP147254
0.1mL Each for $309.05
NBP147254SS Novus Biologicals Canada
NBP147254SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
ERR alpha/NR3B1
Affinity Purified
RUO
Human and mouse. Predicted to react with rat, pig and dog based on 100% sequence homology.
2101
1mg/mL
Synthetic peptide made to an N-terminal portion of human Estrogen Related Receptor alpha (within residues 1-50). [Swiss-Prot No. P11474]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
P11474
PBS, 30% glycerol