missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ERR alpha/NR3B1, Clone: H5844, R&D Systems

$512.30 - $512.30

Specifications

Antigen ERR alpha/NR3B1
Clone H5844
Cross Reactivity Human
Format Purified
Host Species Murine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
PPH584400 R & D Systems
PPH584400
Unlabeled 100μg Each for $512.30
Specifications

Description

Applications:

Direct ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Description & Specifications

Specifications

ERR alpha/NR3B1
Human
Murine
IgG2a
Primary
H5844
Purified
RUO
Monoclonal
Protein A or G purified from hybridoma culture supernatant