Please login to your online account to display your discounted pricing

anti-ERO1L, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7]
Antigen ERO1L
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002525

 
novus biologicals canada
NB1002525
Unlabeled 0.1mL Each for $329.00

NB1002525SS

 
novus biologicals canada
NB1002525SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7]
Antigen ERO1L
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q96HE7
Gene ID (Entrez) 30001
Formulation Borate buffered saline, 1% BSA
Concentration 2mg/mL