Please login to your online account to display your discounted pricing

anti-ERK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056376

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056376

  • / Each
 

Description & Specifications

Specifications

Antigen ERK1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_001103361
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human ERK1 between amino acids 350-400.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5595
Target Species Human